2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%
Acros Organics
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%
CAS: 15871-57-5 | C10H19N2O3 | 215.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR301452500250 mgGlass bottle
Catalog number ACR301452500
Price (MYR)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid
Name Note96%
CAS15871-57-5
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning