4-Fluoro-2-methylphenol, 98%, Thermo Scientific™
4-Fluoro-2-methylphenol, 98%, Thermo Scientific™
4-Fluoro-2-methylphenol, 98%, Thermo Scientific™
Acros Organics
4-Fluoro-2-methylphenol, 98%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR2943700101 gGlass bottle
Catalog number ACR294370010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Fluoro-2-methylphenol
Name Note98%
CAS452-72-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only