▸
Organic compounds
▸
Aryl halides
▸
Aryl chlorides
▸
3-Chlorothiophene, 96%, Thermo Scientific™
Element Search
Structure Search
Acros Organics
3-Chlorothiophene, 96%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
Packaging:
Glass bottle
Catalog number
ACR258800050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3-Chlorothiophene
Name Note
98%
CAS
17249-80-8
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Danger