2-Nitrobenzoyl chloride, 95%
2-Nitrobenzoyl chloride, 95%
2-Nitrobenzoyl chloride, 95%
Acros Organics
2-Nitrobenzoyl chloride, 95%
CAS: 610-14-0 | C7H4ClNO3 | 185.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR232310050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Nitrobenzoyl chloride
CAS610-14-0
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0