N-Benzoyl-L-tyrosine ethyl ester, 98%
N-Benzoyl-L-tyrosine ethyl ester, 98%
N-Benzoyl-L-tyrosine ethyl ester, 98%
Acros Organics
N-Benzoyl-L-tyrosine ethyl ester, 98%
CAS: 3483-82-7 | C18H19NO4 | 313.35 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2275600505 gGlass bottle
Catalog number ACR227560050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Benzoyl-L-tyrosine ethyl ester
Name Note98%
CAS3483-82-7
CAS Min %97.5
CAS Max %100.0