N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
Acros Organics
N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
CAS: 1001-53-2 | Molecular Formula: C4H10N2O | Molecular Weight: 102.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR22317025025 gGlass bottle
Catalog number ACR223170250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-(2-Aminoethyl)acetamide
CAS1001-53-2
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0