Octyltrichlorosilane, 97%
Octyltrichlorosilane, 97%
Octyltrichlorosilane, 97%
Thermo Scientific Chemicals
Octyltrichlorosilane, 97%
CAS: 5283-66-9 | C8H17Cl3Si | 247.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR216501000
Price (MYR)
648.00
Bottle
Quantity:
100 g
Request bulk or custom format
Price (MYR)
648.00
Bottle
Specifications
Chemical Name or MaterialOctyltrichlorosilane
CAS5283-66-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0