Dowex™ 1X8, 100-200 mesh, ion-exchange resin
Dowex™ 1X8, 100-200 mesh, ion-exchange resin
Dowex™ 1X8, 100-200 mesh, ion-exchange resin
Acros Organics
Dowex™ 1X8, 100-200 mesh, ion-exchange resin
CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
Catalog number ACR203001000
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 1X8
Physical FormBeads
CAS12627-85-9
Particle Size15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh)
PackagingPlastic bottle
View more
Ion exchange resin
RUO – Research Use Only