Poly(caprolactone) triol, average M.W. 300
Acros Organics
Poly(caprolactone) triol, average M.W. 300
CAS: 39280-09-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Glass bottle
Catalog number ACR190730250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(caprolactone) triol
ColorColorless
CAS39280-09-6
Physical FormViscous Liquid
MDL NumberMFCD00084405