Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
Acros Organics
Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Packaging:
Glass bottle
Catalog number ACR178060500
Price (MYR)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(acrylic acid)
ColorWhite
CAS9003-01-4
Physical FormPowder
RTECS NumberAT4680000