2-(Bromomethyl)naphthalene, 96%
2-(Bromomethyl)naphthalene, 96%
2-(Bromomethyl)naphthalene, 96%
Acros Organics
2-(Bromomethyl)naphthalene, 96%
CAS: 939-26-4 | C11H9Br | 221.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1625600101 gGlass bottle
Catalog number ACR162560010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-(Bromomethyl)naphthalene
CAS939-26-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0