4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
Thermo Scientific Chemicals
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
CAS: 116-63-2 | C10H9NO4S | 239.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR149270250
Price (MYR)
479.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
479.00
Each
Specifications
Chemical Name or Material4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Name Note95%
CAS116-63-2
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning