Thiocarbanilide, 98%
Thiocarbanilide, 98%
Thiocarbanilide, 98%
Acros Organics
Thiocarbanilide, 98%
CAS: 102-08-9 | C13H12N2S | 228.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR138705000
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThiocarbanilide
Name Note98%
CAS102-08-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.