(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
Acros Organics
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR13373025025 g
Catalog number ACR133730250
Price (MYR)
1,077.00
Bottle
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,077.00
Bottle
Specifications
Chemical Name or Material(±)-erythro-Aleuritic acid
Name Note95%
CAS533-87-9
CAS Min %94.0
CAS Max %100.0