5-Sulfosalicylic Acid Dihydrate, 98%
5-Sulfosalicylic Acid Dihydrate, 98%
5-Sulfosalicylic Acid Dihydrate, 98%
5-Sulfosalicylic Acid Dihydrate, 98%
Acros Organics
5-Sulfosalicylic Acid Dihydrate, 98%
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1328600252.5 kgPlastic bottle
Catalog number ACR132860025
Price (MYR)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5965-83-3, 97-05-2
Specifications
Chemical Name or Material5-Sulfosalicylic acid dihydrate
CAS5965-83-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Indicator
RUO – Research Use Only