L-Phenylalanine methyl ester hydrochloride, 98%
L-Phenylalanine methyl ester hydrochloride, 98%
L-Phenylalanine methyl ester hydrochloride, 98%
Acros Organics
L-Phenylalanine methyl ester hydrochloride, 98%
CAS: 7524-50-7 | C10H14ClNO2 | 215.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number ACR130320025
Price (MYR)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Phenylalanine methyl ester hydrochloride
Name Note98%
CAS7524-50-7
Melting Point156.0°C to 160.0°C