Iodobenzene, 98%
Iodobenzene, 98%
Iodobenzene, 98%
Acros Organics
Iodobenzene, 98%
CAS: 591-50-4 | C6H5I | 204.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1223300252.5 LGlass bottle
Catalog number ACR122330025
Price (MYR)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIodobenzene
CAS591-50-4
Health Hazard 1GHS Signal Word: Warning
Melting Point-29.0°C
ColorYellow