Glycol sulfite, 98%
Glycol sulfite, 98%
Glycol sulfite, 98%
Acros Organics
Glycol sulfite, 98%
CAS: 3741-38-6 | C2H4O3S | 108.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR120120250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycol sulfite
Name Note98%
CAS3741-38-6
ColorColorless
Density1.4200g/mL