Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Organic compounds
▸
Organic oxoazanium compounds
▸
3,4-Dichloronitrobenzene, 95%, Thermo Scientific™
Element Search
Structure Search
Acros Organics
3,4-Dichloronitrobenzene, 95%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number
ACR113530010
Price (MYR)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3, 4-Dichloronitrobenzene
Name Note
95%
CAS
99-54-7
CAS Min %
94.0
Health Hazard 1
GHS Signal Word:
Danger