Thermo Scientific Chemicals
gamma-Butyrolactone, 99+
CAS: 96-48-0 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR108130025
Price (MYR)
1,160.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,160.00
Each
Specifications
Chemical Name or Materialγ-Butyrolactone
CAS96-48-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Health Hazard 3GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.