4-Aminophenyl sulfone, 97%
4-Aminophenyl sulfone, 97%
4-Aminophenyl sulfone, 97%
Acros Organics
4-Aminophenyl sulfone, 97%
CAS: 80-08-0 | C12H12N2O2S | 248.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number ACR104360050
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminophenyl sulfone
Name Note97%
CAS80-08-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning