▸
Acids
▸
Inorganic Acids
▸
Sulfuric Acid
▸
Sulfuric Acid (Optima™), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sulfuric Acid (Optima™), Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
Packaging:
FEP Bottle
Catalog number
A468-1
Price (MYR)
4,495.05
Each
Add to cart
Quantity:
1 L
Packaging:
FEP Bottle
Request bulk or custom format
Price (MYR)
4,495.05
Each
Add to cart
Specifications
Chemical Name or Material
Sulfuric Acid
pH
0.3
Cleanup Symbol
Acid
Color
Colorless to Brown
CAS
7664-93-9
View more
Certificate of Lot Analysis included with each bottle.
Ideal for analysis by ICP-MS and high-resolution ICP-MS.
Evaluated for 65 metals at low-level ppt concentrations.
Manufactured, purified, and packaged in a Class 1000 clean room.
Packaged in specially cleaned Teflon FEP for PFA bottles to maintainproduct integrity and to eliminate metal leaching from glass bottles.
Three year shelf life from time of manufacture available on COA.