S1 Pipet Fillers
Actual product may vary
Thermo Scientific
S1 Pipet Fillers
Carry out longer, fatigue-free pipetting with this ergonomically designed pipet filler, for use with graduated and volumetric glass and plastic serological pipets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDispensing Speed
9501White8
9521Blue8
9531Red8
9541Green, Green8
9511Clear8
Catalog number 9501
HSN 84798999
Price (MYR)
3,498.00
Case
Color:
White
Dispensing Speed:
8
Price (MYR)
3,498.00
Case
Carry out longer, fatigue-free pipetting with the Thermo Scientific™ S1 Pipette Fillers. These instruments are ergonomically designed and offer simple, efficient pipetting performance, as well as maximum pipetting comfort. Lightweight and cordless, they feature a heavy-duty rechargeable lithium-ion battery that allows approximately three times longer operation between charges than typical units. For use with graduated and volumetric glass and plastic serological pipets.
Monitor Performance
  • Large, backlit LCD display provides visual confirmation of remaining battery charge and speed settings
Control Speed
  • Separate aspirate and dispense speed controls provide precise control over pipetting speeds and reduce risk of overpipetting with smaller pipets
  • Choose from eight speeds with a simple “+” or “-” to adjust pipetting speed which is displayed on the LCD
Ready When You Need It
  • Rechargeable lithium-ion battery offers long runtime per charge while eliminating battery memory problems
  • Battery has rapid charge rate of 80% in one hour and delivers up to 15 hours of continuous operation before recharge
Specifications
Battery TypeLithium-Ion
Capacity (Metric)10 to 15 mL
MaterialAR™ Soda Glass
For Use WithCentrifuge
ColorWhite
Dispensing Speed8
No. per Pack100
For Use WithGraduated and Volumetric Glass or Plastic Serological Pipette
ModelS1
Volume (Metric)1 to 100 mL
Filter Type0.45 Micron Standard (Mfr part numbers 4580560 and 9057)
For Use With (Equipment)All major brands of glass or plastic serological pipets, 1 to 100mL
Length (Metric)98 mm
IncludesS1 Pipet Filler, universal charger, table stand, wall-mounted holder, 1mL pipet support, instruction manual and warranty card
Bottom ShapeRound
Recharge Time3 hours
Diameter (Metric)17 mm
GraduatedNo
Product TypeCentrifuge Tube
Warranty2 Years with web registration
Electrical Requirements100 to 240V 50/60Hz
Voltage100/240 V
Amperage0.2A