Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Actual product may vary
Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for four inner-door, ultra-low temperature freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsProduct TypeFor Use With (Equipment)
920090Boxes (2 in.)Sliding Drawer Rack (Tube Boxes)TDE and TSX Series (4 shelves)
1950726Boxes (2 in.)Sliding Drawer Rack (Tube Boxes)Forma 900, Forma 7000, TSE, TSD Sereis (28 cu.ft.)
920091Boxes (3 in.)Sliding Drawer Rack (Tube Boxes)TDE and TSX Series (4 shelves)
Catalog number 920090
HSN 84189900
Price (MYR)
874.00
Each
Holds:
Boxes (2 in.)
Product Type:
Sliding Drawer Rack (Tube Boxes)
For Use With (Equipment):
TDE and TSX Series (4 shelves)
Price (MYR)
874.00
Each
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 4 shelf Series Freezers as well as well as previous generation Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
  • Warranty varies by country, please contact us for details
Specifications
Description2 in. Box Rack - Sliding Drawer
ColorSilver
HoldsBoxes (2 in.)
MaterialStainless Steel
Positions25 Boxes/Rack
ShapeSquare
For Use With (Equipment)TDE and TSX Series (4 shelves)
Width (English)5.5 in.
Height (English)11.9 in.
Depth (English)25.8 in.
Depth (Metric)65.5 cm
Product TypeSliding Drawer Rack (Tube Boxes)
Height (Metric)30.2 cm
Width (Metric)14 cm
Unit SizeEach