Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. Needles are 304 stainless steel (sst.). Standard needle length is 2 in. (51mm). Custom needle lengths and point styles are available.
Have Questions?
Catalog Number
Needle Gauge
For Use With (Equipment)
Autoclavable
90111
11 G
LT and TLL Syringes and Connectors
Not Autoclavable
Catalog number 90111
Price (MYR)
-
Needle Gauge:
11 G
For Use With (Equipment):
LT and TLL Syringes and Connectors
Autoclavable:
Not Autoclavable
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. Needles are 304 stainless steel (sst.). Standard needle length is 2 in. (51mm). Custom needle lengths and point styles are available.
Metal Hub Compatible Syringe Terminations
PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
Needle Point Styles
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 33-10
Six needles per pack.
Specifications
AutoclavableNot Autoclavable
MaterialStainless Steel
Needle Gauge11 G
Length (English) Needle2 in.
Product TypeReplacement Needle
Length (Metric) Needle51 mm
Needle Point StyleSharp, Bevel, Curved
For Use With (Equipment)LT and TLL Syringes and Connectors