Specifications
ColorTan
SizeLarge
For Use With (Application)Food Processing,Picking/Packing,Assembly/Fabrication
HandLeft
Description15mL flat-bottom tube block
Capacity (Metric)15 mL
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds15 x 15mL flat-bottom
LightingLED
No. of Places15
TypeBlock
Dimensions (L x W x H)76 x 124 x 113 mm (3 x 4.9 x 4.55 in.)
Unit SizeEach