Block, 6 mm and 10 mm dia.
Actual product may vary
Thermo Scientific
Block, 6 mm and 10 mm dia.
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog NumberDimensions (L x W x H)
8887011276 x 124 x 39 mm
Catalog number 88870112
HSN 84199090
Price (MYR)
938.00
Each
Dimensions (L x W x H):
76 x 124 x 39 mm
Price (MYR)
938.00
Each
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
DescriptionMixed block, 6mm and 10mm dia.
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds32 x 6mm and 21 x 10mm
LightingLED
TypeBlock
Dimensions (L x W x H)76 x 124 x 39 mm
Unit SizeEach