Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Actual product may vary
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Thermo Scientific
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Speed up your analysis with high-throughput measurements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeHolds
840-314700Rect Cell 4-position Carousel4 x 50mm PL Rectangular Cells
840-3034008-position Carousel8 x 10mm Pathlength Cells
840-3036004-position Carousel Base4 x 10mm Pathlength Cells
Catalog number 840-314700
Price (MYR)
3,318.00
EA
Add to cart
Type:
Rect Cell 4-position Carousel
Holds:
4 x 50mm PL Rectangular Cells
Price (MYR)
3,318.00
EA
Add to cart
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.
Specifications
CompatibilityAccommodates four long-path rectangular cells up to 50mm. Please note: Not compatible with low volume cells (< 3.5mL) or any other type of cell holder.
TypeRect Cell 4-position Carousel
For Use With (Equipment)GENESYS 140/150/180, BioMate 160 Spectrophotometers
Holds4 x 50mm PL Rectangular Cells
Unit SizeEA