General Purpose Centrifuge Rotor Packages
Actual product may vary
General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Have Questions?
Catalog NumberModel
75016105TX-1000 Cell Culture Package
Catalog number 75016105
HSN 84219100
Price (MYR)
31,404.00
Each
Model:
TX-1000 Cell Culture Package
Price (MYR)
31,404.00
Each
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
MaterialSee individual centrifuge, rotor and accessory data
TypeRotor Equipment Package
CapacityMax in TX-1000: 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
Net WeightSee individual components of package
For Use With (Equipment)Purchase of Thermo Scientific General Purpose Centrifuge required for operation
Certifications/ComplianceISO 14971, ISO 13485, ISO 9001
IncludesTX-1000 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters
WarrantySee Manual
BiocontainmentYes
ModelTX-1000 Cell Culture Package
Unit SizeEach