Nalgene™ Lab Notebooks with Regular Paper Pages
Nalgene™ Lab Notebooks with Regular Paper Pages
Actual product may vary
Nalgene™ Lab Notebooks with Regular Paper Pages
Nalgene™ Lab Notebooks with Regular Paper Pages
Nalgene™ Lab Notebooks with Regular Paper Pages
Nalgene™ Lab Notebooks with Regular Paper Pages
Record necessary documentation for securing patent protection. These lab notebooks feature 96 permanently sewn-in paper pages with either grid or horizontal ruled lines.
Have Questions?
Catalog NumberSizeCover ColorRuled
6301-4000A4, 5mm LinesBurgundyYes
Catalog number 6301-4000
HSN 48201090
Price (MYR)
1,184.00
Case
Add to cart
Size:
A4, 5mm Lines
Cover Color:
Burgundy
Ruled:
Yes
Price (MYR)
1,184.00
Case
Add to cart
Lab notebooks include 96 permanently sewn-in paper pages with either grid or horizontal ruled lines. Record necessary documentation for securing patent protection. Pages include spaces for dates and signatures. Notebooks include instructions for keeping records, table of contents and a page for issuing contents. The hard cover is waterproof (pages are not).
Features:
  • Spaces for dates, signature of worker and witness for patent protection
  • Choose forest green cover with 0.25 in. (6mm) green grid or burgundy cover with horizontal lines
  • Page size: Letter (8.5 × 11 in.) or A4 (8.75 × 11.25 in.)
  • Permanently bound and paginated for data security and traceability
Includes:
Instructions for keeping records, page for issuing book and table of contents.
Note:
Pages are not waterproof.
.
.
Specifications
RuledYes
Width (English) Page11.25 in.
Length (English) Page8.75 in.
Cover ColorBurgundy
Cover MaterialPolyethylene
DescriptionLab Notebook Paper
Length (Metric) Page222 mm
Width (Metric) Page285.7 mm
SizeA4, 5mm Lines
Unit SizeCase