CryoBoxes™
Actual product may vary
CryoBoxes™
CryoBoxes™
CryoBoxes™
Thermo Scientific
CryoBoxes™
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHoldsMaterial
5026-0909White81 (1.2 and 2 mL Vials)Polycarbonate
378247White25 (1 to 1.8 mL Vials)Cardboard
5025-0505White25 (1.2 and 2 mL Vials)Polycarbonate
FIS15-350-107BWhite81 (1.2 and 2 mL Vials)Polycarbonate
FKS10324741White81 (1.2 and 2 mL Vials)Polycarbonate
5026-1010Blue100 (1 to 1.5 mL Vials)Polycarbonate
867013-0243Blue81 (1 to 2 mL Vials)Polycarbonate
Catalog number 5026-0909
HSN 39231090
Price (MYR)
405.00
PK
Color:
White
Holds:
81 (1.2 and 2 mL Vials)
Material:
Polycarbonate
Price (MYR)
405.00
PK
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes. Choose from a variety of durable plastic Thermo Scientific™ CryoBox™ styles with keyed lids and printed location grids, or select economical cardboard/chipboard CryoBoxes™ for short-term storage. The plastic boxes are optimized to fit our own brand of Nalgene™ and Nunc™ vials and microcentrifuge tubes best.
  • Durable, all-plastic CryoBoxes™ for short- or long-term storage
  • Keyed lids on plastic CryoBoxes™ prevent misalignment
  • Plastic boxes are autoclavable for decontamination and usable at temperature range from -196°C to +121°C
  • Printed grid on plastic lids for easy inventory management
  • Economical chipboard/cardboard CryoBoxes™ for short term storage
  • Chipboard/cardboard boxes have dividers and vents in base for good drainage and air circulation
  • Available with dividers from 25 to 169 tube storage
Specifications
DescriptionPolycarbonate, 133 x 133 x 52mm
ColorWhite
MaterialPolycarbonate
ShapeSquare
TypeCryo Box
Holds81 (1.2 and 2 mL Vials)
Product LineNalgene
Quantity4/Pk., 24/Cs.
Tube Array9 x 9
Dimensions (L x W x H)5.25 x 5.25 x 2 in.
Length (English)5.25 in.
Length (Metric)133 mm
Width (English)5.25 in.
Width (Metric)133 mm
Height (English)2 in.
Height (Metric)52 mm
Weight (Metric)4 kg
Unit SizePK