Point of Dispense Filters for Barnstead™ Point of Use Water Purification Systems
Actual product may vary
Point of Dispense Filters for Barnstead™ Point of Use Water Purification Systems
Sterile filter for use with Barnstead™ water purification systems; can be autoclaved up to 5 times.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
50157375Sterile Filter, 0.1μm
09.1003Sterile Filter, 0.2μm
Catalog number 50157375
HSN 84212190
Price (MYR)
1,652.00
Each
Description:
Sterile Filter, 0.1μm
Price (MYR)
1,652.00
Each
Reduce bacteria using the Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems.
Replacement consumable for the following Barnstead water purification systems:
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • LabTower™ EDI
  • LabTower™ TII
  • MicroPure™
  • Smart2Pure™
  • Smart2Pure™ Pro
Specifications
Cartridge TypePoint of Dispense filter
DescriptionSterile Filter, 0.1μm
For Use With (Equipment)Smart2Pure Pro
Product TypeWater Purification System
Unit SizeEach