Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Have Questions?
Change view
Catalog Number
Type
50134022
Filter
09.4000
Pretreatment Cartridge
06.5204
Filter with Stabilizer
06.5452
Stabilizing Cartridge
09.4003
Pretreatment Filter
5 Options
Catalog number 50134022
HSN 84212190
Price (MYR)
926.00
Each
Type:
Filter
Price (MYR)
926.00
Each
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
For Use With (Equipment)LabTower EDI, LabTower RO, LabTower TII
TypeFilter
Includes5 &um; Filter w/ Carbon, 10 ft. Filter Housing