FluidEase™ Pro ClipTip™ Electronic Multichannel Pipettes
Actual product may vary
FluidEase™ Pro ClipTip™ Electronic Multichannel Pipettes
Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic multichannel pipettes enable effortless 96- and 384-well plate filling with touchscreen control, ultra-wide speed selection, and secure ClipTip™ sealing for reproducible, high-throughput workflows in shared labs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Number of Channels
46761201 to 30 μL16
46760100.5 to 10 μL8
46760302.5 to 100 μL8
46760505 to 200 μL8
467607010 to 300 μL8
467609025 to 1250 μL8
46760200.5 to 10 μL12
46760402.5 to 100 μL12
46760605 to 200 μL12
467608010 to 300 μL12
467610025 to 1250 μL12
46761100.5 to 10 μL16
46761303 to 125 μL16
Catalog number 4676120
HSN 84189900
Price (MYR)
18,744.00
Each
Volume (Metric):
1 to 30 μL
Number of Channels:
16
Price (MYR)
18,744.00
Each
Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic multichannel pipettes are designed to simplify and standardize plate-based workflows. A large touchscreen interface and ultra-wide speed selection support precise aspiration and dispense across liquid types. Secure ClipTip™ sealing, effortless column-to-row transitions, preset and advanced programming modes, and password-protected settings enable reproducible, high-throughput plate filling across shared laboratory environments
Features:
  • Large Touchscreen User Interface: A high-resolution touchscreen provides direct, visual interaction for setup and execution, reducing training time and minimizing setup errors compared to button- or joystick-based navigation.
  • Ultra-Wide Speed Selection: One of the broadest aspiration and dispense speed ranges available supports gentle handling of sensitive samples, precise control of viscous liquids, and faster execution for time-critical workflows.
  • Simple Presets and Advanced Programming: Easy-to-use preset modes support fast onboarding for new or occasional users, while advanced step programming enables complex, multi-step protocols for demanding applications.
  • Index-Finger Operation with Increased Rotational Flexibility: Retains Thermo Fisher–recognized index-finger operation with a wider angle of rotation at the finger rest to accommodate different hand sizes, grip angles, and working positions.
  • Secure ClipTip™ Technology: ClipTip™ tips lock and seal in place until intentional ejection, helping ensure consistent volume delivery and reduced variability from dispense to dispense.
  • Electronic Tip Ejection with Safe-to-Eject Illumination: Electronic tip ejection reduces physical effort and includes LED illumination to indicate when ejection is possible, providing ergonomic benefit and added user security.
  • Password Protection and User Profiles: Password-protect critical settings and protocols and support up to five user profiles to enable standardized operation in shared and regulated laboratory environments.
  • Custom Calibration Options: Enable calibration to customer-specific liquids or environmental conditions to align performance with real laboratory workflows.
  • Service and Maintenance Tracking: Built-in service reminders, calibration logs, and usage tracking support instrument management and compliance documentation over time.
Specifications
Adjustable Tip SpacingNo
AutoclavableNot Autoclavable
ColorViolet
DisposableReusable
For Use WithClipTip 384 30
ModelBluetooth
Number of Channels16
ProgrammabilityYes
SterilityNon-sterile
TypeElectronic Pipette
Volume (Metric)1 to 30 μL
DisplayTouchscreen
Finger StyleErgonomic Index Finger
Multilingual User InterfaceYes
Warranty1 Year, 2 Years with Registration
Unit SizeEach