Specifications
Adjustable Tip SpacingNo
AutoclavableNot Autoclavable
ColorPink
DisposableReusable
For Use WithClipTip 12.5
ModelBluetooth
Number of Channels8
ProgrammabilityYes
SterilityNon-sterile
TypeElectronic Pipette
Volume (Metric)0.5 to 10 μL
DisplayTouchscreen
Finger StyleErgonomic Index Finger
Multilingual User InterfaceYes
Warranty1 Year, 2 Years with Registration
Unit SizeEach