Finnpipette Stepper Pipette
Actual product may vary
Thermo Scientific
Finnpipette Stepper Pipette
Rapidly dispense up to 45 times in succession without refilling. This lightweight, easy to use repeater pipetter is designed for one-handed dispensing.
Have Questions?
Catalog NumberVolume (Metric)
454000010 to 5000 μL
Catalog number 4540000
HSN 84798999
Price (MYR)
-
Volume (Metric):
10 to 5000 μL
Rapidly dispense up to 45 times in succession without refilling. The Thermo Scientific™ Finnpipette™ Stepper Pipette is a lightweight, easy to use repeater pipette ergonomically designed for one-handed dispensing. Operating on the positive displacement principle, the Finnpipette Stepper is ideally suited for work with aggressive and viscous liquids.
Features:
  • Positive displacement design ensures accurate results, even for viscous, volatile, or other problematic liquids
  • Lightweight, ergonomic handle and wide finger rest minimize operator hand strain
  • Operation is simple—just attach the appropriate tip size, fill and prime the tip, and dial the selected volume
  • Volume is easy to set, with the aid of the convenient volume-setting chart located on the handle
Ordering Information:
Universal Stepper handle is also available with an eight-channel module. For convenient storage, pipetter stand is available (Cat. No. 9420290).
Includes:
Unit comes with one adapter (nonsterile) necessary for use with 25 or 50mL tips.
Requires:
Uses the Thermo Scientific Finntip™ Stepper tip, a plunger-style tip available in nonsterile or sterile (individually wrapped) form. This tip comes in seven sizes from 0.5 to 50mL, each with five different delivery volumes, for an overall dispensing range of 10 to 5000μL. Only the disposable tip comes in contact with the sample, eliminating the risk of contaminating the pipetter body.
Specifications
DescriptionFinnpipette Stepper Pipetter
For Use WithFinnTip
ModelStepper
Number of Channels1
TypeStepper Pipette
Volume (Metric)10 to 5000 μL
Unit SizeEach