Hypersil™ APS-2 Amino HPLC Columns
Actual product may vary
Thermo Scientific
Hypersil™ APS-2 Amino HPLC Columns
Analyze sugars with this versatile amino propyl phase of Thermo Scientific™ Hypersil™ APS-2 Amino LC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
30705-254630250 mm4.6 mm5 μm
HYP30705-254630250 mm4.6 mm5 μm
30705-154630150 mm4.6 mm5 μm
HYP30705-154630150 mm4.6 mm5 μm
30703-01400110 mm4 mm3 μm
Catalog number 30705-254630
HSN 90279090
Price (MYR)
-
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
Analyze sugars with this versatile amino propyl phase of Thermo Scientific™ Hypersil™ APS-2 Amino LC Columns. Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines. For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ LC Amino Columns as an alternative.
  • In reversed phase mode, Hypersil APS-2 columns are excellent for carbohydrate analysis: extra sensitivity for sugars
  • Alternative selectivity to silica when used in normal phase
Ordering Information:
Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithIC
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size120 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL8
Stationary PhaseAmino (NH2)
Product LineHypersil Classical
Packing MaterialSpherical, Fully Porous Type A Silica
Carbon Load2%
Column TypeMixed Mode
EndcappedNo
PhaseReversed Phase, Normal Phase, Ion Exchange, HILIC
Product TypeAnalytical Column
Unit SizeEach