Search Fisher Scientific
| Catalog Number | Product Type | Holds | For Use With (Equipment) |
|---|---|---|---|
| 30160 | Flask Clamp | 5 L Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30129 | Flask Clamp Kit | Flasks | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30152BI | Flask Clamp | 50 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30154BI | Flask Clamp | 250 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30155 | Flask Clamp | 300 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30156BI | Flask Clamp | 500 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30157BI | Flask Clamp | 1 L Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30158 | Flask Clamp | 2 L Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30175 | Microplate/Deep-Well Plate Clamp | Microplate/Deep-Well Plate | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
| 30175DBQ | Microplate/Deep-Well Plate Clamp | Microplate/Deep-Well Plate | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.