Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Actual product may vary
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Thermo Scientific
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.
Have Questions?
Catalog NumberCapacity (Metric)
2123-00104 L
Catalog number 2123-0010
HSN 39233090
Price (MYR)
1,170.00
Case
Capacity (Metric):
4 L
Price (MYR)
1,170.00
Case
Collect, store and transport dry chemicals and environmental samples with Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure. Molded-in handgrips provide greater convenience and safety when pouring.
  • Useful for collecting, storing and transporting dry chemicals, environmental samples, reagents and distilled water
  • Good for freezing to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols
  • Square shape saves space, handles easily
  • Wide mouth gives easy access
  • Polypropylene cap included
  • Not liquid leakproof
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Specifications
Capacity (Metric)4 L
Capacity (English)1 gal.
ColorTranslucent
DescriptionLarge Wide-Mouth Square Bottle
Diameter (English)5.6 in.
Diameter (English) Inner Neck3.5 in.
Diameter (Metric) Inner Neck89 mm
MaterialHDPE
No. per Pack1
SterilityNon-sterile
Closure ColorWhite
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
GraduatedNo
MouthWide
ShapeSquare
Height (Metric)292 mm
Product TypeWide Mouth Bottle (Large)
Width (English)5.66 in.
Height (English)11.5 in.
Closure MaterialPolypropylene
Closure Size100 mm
Diameter (Metric)142 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCase
Frequently asked questions (FAQs)
Why are some of your wide mouth bottles and jars not considered liquid leakproof?
This was a consequence of design and engineering limitations, and there was no practical way to incorporate the same level of seal integrity as our certified leakproof containers. While in many cases, they still hold up well to liquid containment applications, they have not been tested specifically for that purpose, and thus would be the responsibility of the customer to perform validation and risk management studies if necessary.