Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure
Actual product may vary
Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure
Thermo Scientific
Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure
Easily measure and dispense using these translucent, square, narrow-mouth PPCO bottles, featuring mouths wider than conventional narrow-mouth bottles.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
2016-006060 mL24 mm
Catalog number 2016-0060
HSN 39233090
Price (MYR)
943.00
Case
Capacity (Metric):
60 mL
Closure Size:
24 mm
Price (MYR)
943.00
Case
Easily measure and dispense using Thermo Scientific™ Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure, featuring mouths wider than conventional narrow-mouth bottles. These polypropylene copolymer bottles are graduated, repeatedly autoclavable, translucent, and resistant to alcohols and bases. They are made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.
Features:
  • Square sides allow for good use of space and an easy grip
  • Graduated for ease of dispensing
  • Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable/Leakproof† when used with Nalgene closures
  • Translucent bottles provide visible liquid level
  • Useful between 0° and 135°C (32° and 275°F)
  • Manufactured from non-toxic materials
Includes:
  • Leakproof† polypropylene screw caps
Note:
Remove cap or completely disengage threads before autoclaving. Bottle neck design is protected by US Trademark Reg. No. 2857283. Square bottle with arched shoulders design is protected by US Trademark Reg. No. 2857279.
.
Recycling Information:
Nalgene reusable polypropylene (PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity2 oz., 60 mL
Capacity (Metric)60 mL
ColorTranslucent
DescriptionNarrow-Mouth Square Bottle
Diameter (English)1.6 in.
Diameter (English) Inner Neck0.7 in.
Diameter (Metric) Inner Neck18 mm
IncludesLeakproof polypropylene screw closures
MaterialPPCO
No. per Case72
No. per Pack12
SterilityNon-sterile
Closure ColorNatural, translucent
GraduationsYes
Closure IncludedYes
GraduatedYes
MouthNarrow
Height (Metric)83 mm
Width (English)1.61 in.
Height (English)3.3 in.
Closure MaterialPolypropylene
Closure Size24 mm
Diameter (Metric)41 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
AutoclavableAutoclavable
Capacity (English)2 oz.
ShapeSquare
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Product TypeSquare Narrow Mouth Bottle
Unit SizeCase
Frequently asked questions (FAQs)
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.