accumet™ Glass Body Ag/AgCl Reference Electrodes - Mercury-Free
Actual product may vary
Thermo Scientific
accumet™ Glass Body Ag/AgCl Reference Electrodes - Mercury-Free
Recommended for stability at higher temperatures and for use with strongly basic samples
Have Questions?
Change viewbuttonViewtableView
Catalog NumberReference Electrolyte
13620273SP138-500, SP135-500
FSP13-620-53SP138-500
FSP13-620-273SP138-500, SP135-500
FIS13-620-21613-620-833
FSP13-620-21613-620-833
Catalog number 13620273
HSN 90279090
Price (MYR)
-
Reference Electrolyte:
SP138-500, SP135-500
Recommended for stability at higher temperatures and for use with strongly basic samples
Glass body for resistance to wide variety of solutions
  • Operating temperature range of 0° to +110°C (32° to 212°F)
  • 0 to 14pH range
  • Standard reference pin connector
  • All have 8μL/hr. flowrate
Single-Junction Standard Model
  • Standard size glass body
  • Prefilled with 4M KCl saturated with AgCl
  • Stable at higher temperatures and with strongly basic samples
Single-Junction Side Arm Model
  • Standard size glass body with 1 in. (25mm) tubulation fitting, handy for procedures that require remote filling
  • Prefilled with 4M KCl saturated with AgCl
  • Stable at higher temperatures and with strongly basic samples
Single-Junction Silver/Chloride Titration Model
  • Standard size glass body
  • Prefilled with 1M sodium sulfate for silver and chloride titrations
  • Stable at higher temperatures and with strongly basic samples
Double-Junction Model
  • Designed for ISE and sensitive pH measurements, titrations and ORP testing
  • Inner body is prefilled with 4M KCl saturated with AgCl
  • Outer body is packed without electrolyte, so you can choose a compatible electrolyte for your samples
  • Flowrate, 8μL/hr.
  • U.S. Patent No. 4,390,406
Specifications
Connector TypesPin
JunctionDouble
MaterialGlass
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypeAg/AgCl Reference Electrode
ElectrodeSuggested Indicating Electrodes: 13-620-122, 13-620-520, 13-620-521, 13-620-518, 13-620-519, 13-620-500, 13-620-501
Filling Solution Type4mol/L KCl with AgCl
For Use With (Application)Recommended for stability at higher temperatures and for use with strongly basic samples
Internal ReferenceAg/AgCl
ModelDouble-Junction High Temperature Ag/AgCl Reference
Reference ElectrolyteSP138-500, SP135-500
Temperature (English) Operating32°F to 212°F
Temperature (Metric) Range0°C to 100°C
Length (Metric)106 mm
Diameter (Metric) Body12 mm
Unit SizeEach