Pretreatment Cartridges and Filters
Actual product may vary
Pretreatment Cartridges and Filters
Pretreatment Cartridges and Filters
Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
06.5204Filter with Stabilizer
09.4000Pretreatment Cartridge
06.5452Stabilizing Cartridge
09.4003Pretreatment Filter
50134022Filter
Catalog number 06.5204
HSN 84212190
Price (MYR)
1,078.00
Each
Type:
Filter with Stabilizer
Price (MYR)
1,078.00
Each
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
For Use With (Equipment)LabTower EDI, LabTower RO
TypeFilter with Stabilizer
Includes5 μm Filter, Hardness Stabilizer
Unit SizeEach