Check Order Status
Rubber Vacuum Hose
Actual product may vary
Rubber Vacuum Hose
Thick-walled red rubber vacuum hose. Welch Ilmvac™ Rubber Vacuum Hose well suited for laboratory vacuum pump processes.
Have Questions?
Catalog Number
Tubing Size
WEL828310-4
8mm ID, 18mm OD
Catalog number
WEL828310-4
Price (MYR)
430.00
EA
Add to cart
Tubing Size:
8mm ID, 18mm OD
Price (MYR)
430.00
EA
Add to cart
Economical
Flexible
Resilient
Thick-walled
Specifications
Diameter (Metric) Inner
8 mm
Material
Rubber
Diameter (Metric) Outer
18 mm
Overall Length (Metric)
100 cm
Packaging Quantity
1 Each
Tubing Size
8mm ID, 18mm OD
Color
Red
Product Type
Vacuum Hose