Dry, Oil-Free, Two-Stage, Chemical Duty Diaphragm Pump, MPC 095 Z, Ultimate Pressure 5 Mbar
Dry, Oil-Free, Two-Stage, Chemical Duty Diaphragm Pump, MPC 095 Z, Ultimate Pressure 5 Mbar
Actual product may vary
Dry, Oil-Free, Two-Stage, Chemical Duty Diaphragm Pump, MPC 095 Z, Ultimate Pressure 5 Mbar
Welch-Ilmvac™ Dry, Oil-Free, Two-Stage, Chemical Duty Diaphragm Pumps (MPC 095 Z) can deliver an ultimate pressure of 5 Mbar at noise levels of <45 dB. Supplied with a gas ballast valve.
Have Questions?
Catalog number WEL412422-02
Price (MYR)
-
  • Product Type: Diaphragm Pump
  • Oil-Free: Yes
  • Number of Stages: Two
  • Application: Chemical Duty
  • Air Displacement: 15 L/min.
  • Ultimate Pressure: 5 Mbar
  • Noise Level: Less Than 45 dB
  • Dimensions (W/D/H): 235 mm x 140 mm x 277 mm
  • Weight: 6.15 kg
  • Power Supply: 90-260VAC
Specifications
IncludesGas Ballast Valve
Depth (Metric)140 mm
ModelMPC 095 Z
Noise Level<45 dB
Weight (Metric)6.15 kg
Product LineOil-Free Diaphragm Pump
Overall Height (Metric)277 mm
Overall Width (Metric)235 mm
Pump TypeVacuum System
Air Displacement15L/min.
No. of Pump Heads2
Oil CapacityOil-Free
ApplicationsChemical Duty
No. of Stages2
Maximum Pressure5 Mbar
Voltage90/260 VAC
Unit SizeEA