Acid Neutralization Trap for Vacuum Pumps
Acid Neutralization Trap for Vacuum Pumps
Actual product may vary
Acid Neutralization Trap for Vacuum Pumps
Accessory for Welch Vacuum Pump Models 1402N, 1376N
Have Questions?
Catalog NumberSize
WEL1420H-21Up to 2 cfm (57 L/min.)
Catalog number WEL1420H-21
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
Up to 2 cfm (57 L/min.)
For use with Welch Vacuum Pump Models 1402N, 1376N
  • Contains large alkaline element cartridge to neutralize hydrous acids before they enter vacuum pump
  • Transparent body
  • Capacity: Up to 2 cfm and 12 cfm
  • 20.3W x 33cm H
Specifications
For Use WithModel 1402N, 1376N vacuum pump
Overall Height (English)8.97 in.
Overall Height (Metric)22.78 cm
Overall Width (English)7.99 in.
SizeUp to 2 cfm (57 L/min.)
TypeAcid Neutralization Trap
Unit SizeEA