UltraFast™ UFC-WAX GC Columns
UltraFast™ UFC-WAX GC Columns
Actual product may vary
UltraFast™ UFC-WAX GC Columns
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns.
Have Questions?
Catalog NumberFilm Thickness
UFMC000010105010.1 μm
Catalog number UFMC00001010501
Price (MYR)
-
Film Thickness:
0.1 μm
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns. If your laboratory processes large numbers of samples each day, UltraFast GC columns have the processing speed and throughput you need. These short, narrow-bore columns are designed for use with the Thermo Scientific TRACE GC UltraFast gas chromatograph configuration. Together they deliver a significant gain in analysis speed compared to conventional GC procedures.
Applications:
Chemical
Petrochemical
Environmental
Flavors and fragrances
Tailored Specifically for Robustness and Speed
UltraFast GC columns utilize short (2 to 10m), narrow-bore capillaries and temperature programming conditions usually fast than 2°C/s, which results in peak widths in the 50-200ms range. As a result, analysis times average 1 minute or less. Benefits include:
Dramatically shorter analysis times
Increased sample throughput–by a factor of 20
Longer column lifetimes
UltraFast UFC-WAX Columns uses:
Fatty acid methyl esters (FAMEs)
Essential oils
General
Specifications
For Use With (Application)GC for Chemicals, GC for Petrochemicals, GC for Environmental Analysis, GC for Flavors and Fragrances
Length (Metric)5 m
Diameter (Metric) Inner0.1 mm
Stationary PhaseUltraFast UFC-WAX
For Use With (Equipment)Trace GC UltraFast Instrument
Description5 m x 0.1 mm x 0.1 μm
Product LineUltrafast
Film Thickness0.1 μm
Unit SizeEach