Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Actual product may vary
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Have Questions?
Catalog NumberTypeDescriptionSwept Volume
U-435
also known as IDEX = U-435
Union0.010 in. through hole0.025 μL
Catalog number U-435
also known as IDEX = U-435
HSN 90279090
Price (MYR)
0.00
Each
Add to cart
Type:
Union
Description:
0.010 in. through hole
Swept Volume:
0.025 μL
Price (MYR)
0.00
Each
Add to cart
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
  • Absolute zero or low dead volume formats
  • Includes two stainless steel nuts and ferrules
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeUnion
Diameter (English) Inner0.01 in.
Diameter (Metric) Outer0.25 mm
Pressure Rating10000 psi
Swept Volume0.025 μL
IncludesUnion with 2 stainless steel nuts and ferrules
Description0.010 in. through hole
MaterialStainless Steel
For Use With (Equipment)Upchurch Scientific/Parker fittings
Unit SizeEach