Sieve Certification and Recertification Services
Product Image
Sieve Certification and Recertification Services
Certification services offered for new or used sieves
Have Questions?
Catalog NumberProduct Type
TYL9998Certification Service for new sieves
Catalog number TYL9998
Price (MYR)
985.00
EA
Add to cart
Product Type:
Certification Service for new sieves
Price (MYR)
985.00
EA
Add to cart
Certification Service
  • Order when purchasing new sieves
  • Uses a computerized video imaging system to verify that each sieve opening meets ASTM™ and ISO specifications within a tolerance of ±0.1 microns
  • Sieves are then serialized and supplied with documentation, traceable to NIST™, detailing the openings and the wire diameters of the mesh
Recertification Services
  • Offer the same examination and documentation processes for in-use sieves as for new sieves
  • Contact your Fisher Scientific Customer Service Representative for the recommended sieve aperture/recertification schedule
Specifications
Product TypeCertification Service for new sieves
For Use With (Application)To serialize and supply with documentation
Unit SizeEA