Check Order Status
SPE Columns, Drug Abuse (C8 + Benzyl Sulfonic Acid), 200 mg/3 mL
SPE Columns, Drug Abuse (C8 + Benzyl Sulfonic Acid), 200 mg/3 mL
Medical-grade polypropylene with PE frits
Have Questions?
Catalog number
TYL9902
Price (MYR)
347.00
EA
Add to cart
Price (MYR)
347.00
EA
Add to cart
Wide selection of support materials and multiple cartridge/column sizes
Every stationary phase batch tested to ensure lot-to-lot reproducibility
B&J's reliable quality
Specifications
Description
50/Pk.