U.S. Stainless Steel Test Sieve Covers
U.S. Stainless Steel Test Sieve Covers
Actual product may vary
U.S. Stainless Steel Test Sieve Covers
Sieve cover, for U.S. standard test sieve; Stainless Steel; For 8 in. dia. (20.3cm) sieves
Have Questions?
Catalog NumberDiameter (English) Outer
TYL84968 in.
Catalog number TYL8496
Price (MYR)
302.00
EA
Add to cart
Diameter (English) Outer:
8 in.
Price (MYR)
302.00
EA
Add to cart
Cover, Sieve; Fisherbrand; For U.S. standard test sieve; Stainless Steel; For 8 in. dia. (20.3cm) sieves
Note: This is the lid only. For use with Stainless Steel pan (Cat. No. 04-887B).
  • Stainless Steel
  • Fits all 8 and 12 in. diameter (20.3 and 30.5 cm) sieves.
Specifications
TypeSieve Cover
Diameter (English) Outer8 in.
For Use With (Equipment)8 in. dia. Test sieve receiver (04-887B)
Diameter (Metric) Outer203 mm
Frame MaterialStainless Steel
Mesh MaterialStainless Steel
Unit SizeEA